Tisk…

Betulinic acid

Dodavatel: Thermo Scientific

Synonyms: (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-Hydroxy-5a,5b,8,8,11a-pentamethyl-1-(prop-1-en-2-yl)icosahydro-1H-cyclopenta[a]chrysene-3a-carboxylic acid, 3beta-Hydroxy-20(29)-lupaene-28-oic Acid

462870010 462872500
ACRO462870010EA 12950 CZK
ACRO462870010 ACRO462872500
Betulinic acid
Betulinic acid
Vzorec: C₃₀H₄₈O₃
MW: 456,71 g/mol
Bod tání: 295…298 °C
Storage Temperature: Freezer
MDL Number: MFCD00009619
CAS číslo: 472-15-1

Objednat

Výsledky specifikačního testu

Appearance White to light beige Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Solubility (0.1% in ethanol) Clear colorless
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR