Tisk…

4-tert-Butylphenyl glycidyl ether 95%

Dodavatel: Thermo Fisher Scientific
Varování

H55945.06 H55945.14
H55945.06EA 1720 CZK
H55945.06 H55945.14
4-tert-Butylphenyl glycidyl ether 95%
4-tert-Butylphenyl glycidyl ether

Used in paint industry and serves as organic coating in paint, boat paint, building coating material and also as an epoxy reactive diluent.

Vzorec: C₁₃H₁₈O₂
Bod varu: 165…170 °C (14 mmHg)
Hustota: 1,038
Bod vzplanutí: 102 °C (216 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005136
CAS číslo: 3101-60-8
EINECS: 221-453-2

Objednat

Výsledky specifikačního testu

Assay (GC) 94,0%
Refractive index 1,5150
Solubility Slightly soluble in water


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR