Tisk…
Capillary columns, Chromolith® CapRod®
Dodavatel: Merck
1.50402.0001 1.50404.0001 1.50405.0001 1.50407.0001 1.50400.0001 1.50403.0001 1.50409.0001 1.50426.0001 1.50424.0001 1.52031.0001
1.50402.0001EA 38770 CZK
1.50402.0001 1.50404.0001 1.50405.0001 1.50407.0001 1.50400.0001 1.50403.0001 1.50409.0001 1.50426.0001 1.50424.0001 1.52031.0001
Capillary columns, Chromolith® CapRod®
Kolony chromatografické
Fast nano-LC for proteins and peptides. Chromolith® CapRod® capillaries combine the speed of monolithic silica technology with the sensitivity of nano-LC, thus enabling high speed high sensitivity LC-MS analytics.
  • High flow rates at low pressure
  • Long column lifetime
  • Robust handling