Nafcillin sodium salt
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
461380010
461380050
ACRO461380010EA
3170
CZK
ACRO461380010
ACRO461380050
Nafcillin sodium salt
Nafcillin sodium salt
Vzorec:
C₂₁H₂₁N₂NaO₅S Storage Temperature: Refrigerator |
MDL Number:
MFCD00056863 CAS číslo: 985-16-0 |
Výsledky specifikačního testu
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Water | ≤6 % |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...