Tisk…

D(+)-Sacharóza ≥99%, white crystalline powder cell culture reagent

Dodavatel: MP Biomedicals

02194747.5 0219474701 0219474705
ICNA02194747.5EA 1980 CZK
ICNA02194747.5 ICNA0219474701 ICNA0219474705
D(+)-Sacharóza ≥99%, white crystalline powder cell culture reagent
D(+)-Sacharóza

Storage: Store at room Temperature (15-30 °C)
Sucrose is a nonreducing disaccharide composed of glucose and fructose, linked via their anomeric carbons. It is obtained commercially from Sugarcane, sugar beet (Beta vulgaris), and other plants and used extensively as a food and a sweetener.
Use to create sucrose gradients for purification of viruses and proteins.


  • Cell Culture Reagent Grade
  • Partition Coefficient: log Kow = -3.70 (Lit.)
  • pKa: 12.62 (Lit.)

Vzorec: C₁₂H₂₂O₁₁
Bod tání: 177…187 °C
Hustota: 1,587 (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS číslo: 57-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Identity Test Passes
FTIR Conforms to Standard
Purity ≥99%
UV/VIS (30% aq) OD260 ≤0.1
Optical Rotation +66.0°±5 (H2O, NH4OH)
Solubility (50% aq soln) Clear and Colorless
Heavy Metals <0.0005%
RNAse Negative
DNAse Negative


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR