Trimethyl citrate
Dodavatel: Thermo Fisher Scientific
Synonyms:
Citric acid trimethyl ester
462250050
462250250
ACRO462250050EA
1290
CZK
ACRO462250050
ACRO462250250
Trimethyl citrate
Trimethyl citrate
Vzorec:
C₉H₁₄O₇ MW: 234.21 Bod varu: 176 °C (16 mmHg) Bod tání: 75…78 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00025889 CAS číslo: 1587-20-8 |
Výsledky specifikačního testu
Appearance | White to off-white Crystals |
Infrared spectrum | Conforms |
Melting point | 75 to 82 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...