Fluticasone propionate
Dodavatel: Thermo Fisher Scientific
462101000
462105000
ACRO462101000EA
1960
CZK
ACRO462101000
ACRO462105000
Fluticasone propionate
Fluticasone propionate
Vzorec:
C₂₅H₃₁F₃O₅S Bod tání: 272…273 °C Storage Temperature: Freezer |
MDL Number:
MFCD00866007 CAS číslo: 80474-14-2 |
Výsledky specifikačního testu
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % |
HPLC | ≥96.0 % |
Specific optical rotation | +32° to +36° (20°C, 589 nm) (0.5% in CH2Cl2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...