L(-)-Glutathion (redukovaná forma) ≥98%, off-white powder cell culture reagent
Dodavatel: MP Biomedicals
Synonyms:
L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
Storage: Store at +4 °C, store under nitrogen
Glutathione is the major low molecular weight thiol compound of the living plant or animal cell. It is a tripeptide with a gamma peptide linkage between the amine group of cysteine (which is attached by normal peptide linkage to a glycine) and the carboxyl group of the glutamate side-chain. It is an antioxidant, preventing damage to important cellular components caused by reactive oxygen species such as free radicals and peroxides. The sulfhydryl (thiol) group (SH) of cysteine serves as a proton donor and is responsible for the biological activity of glutathione.
Glutathione suppresses human immunodeficiency virus expression in chronically infected monocytic cells. It is a useful tripeptide involved in many aspects of metabolism, including transport of g-glutanyl amino acids and reductive cleavage of disulfide bonds.
Endogenous antioxidant that plays a major role in reducing reactive oxygen species formed during cellular metabolism and the respiratory burst. Glutathione-S-transferase catalyzes the formation of glutathione thioethers with xenobiotics, leukotrienes, and other molecules that have an electrophilic center. Glutathione also forms disulfide bonds with cysteine residues in proteins. Via these mechanisms, it can have the paradoxical effect of reducing the efficacy of anti-cancer agents.
- Cell Culture Reagent Grade
- Presentation: Off-White Powder
- Amino Acid Sequence :γ-Glu-Cys-Gly
- Soluble in water giving clear and colorless solution (3.5% aq soln).
Vzorec:
C₁₀H₁₇N₃O₆S MW: 307,33 g/mol Bod tání: 192…195 °C (lit.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00065939 CAS číslo: 70-18-8 |
Výsledky specifikačního testu
Identity Test A | Passes |
Identity Test B | Passes |
Purity | ≥98% |
FTIR | Conforms to Standard |
Solubility (3.5% aq soln) | Clear, Colorless Solution |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...