Tenofovir disoproxil fumarate 98%
Dodavatel: Thermo Scientific
Synonyms:
(R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene) diisopropyl dicarbonate fumarate
461250010
461250050
461250250
ACRO461250010EA
1300
CZK
ACRO461250010
ACRO461250050
ACRO461250250
Tenofovir disoproxil fumarate 98%
Tenofovir disoproxil fumarate
Vzorec:
C₁₉H₃₀N₅O₁₀P·C₄H₄O₄ MW: 635,5 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08141829 CAS číslo: 202138-50-9 |
Výsledky specifikačního testu
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤1 % |
HPLC | ≥97.5 % |
Fumaric acid | 17.5 to 19 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...