Imatinib mesylate 98%
Dodavatel: Thermo Fisher Scientific
461080010
461080050
ACRO461080010EA
8060
CZK
ACRO461080010
ACRO461080050
Imatinib mesylate 98%
Imatinib mesylate
Vzorec:
C₂₉H₃₁N₇O·CH₄SO₃ Bod tání: 209 °C Storage Temperature: Freezer |
MDL Number:
MFCD04307699 CAS číslo: 220127-57-1 |
Výsledky specifikačního testu
Appearance | White to light yelow or light beige Powder |
Infrared spectrum | Conforms |
Water | ≤1 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...