Tisk…

(S)-(+)-Clopidogrel sulphate 98%

Dodavatel: Thermo Fisher Scientific

461030010 461030050
ACRO461030010EA 3440 CZK
ACRO461030010 ACRO461030050
(S)-(+)-Clopidogrel sulphate 98%
(S)-(+)-Clopidogrel sulphate
Vzorec: C₁₆H₁₆ClNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00876395
CAS číslo: 120202-66-6

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %
Specific optical rotation +54° to +58° (20°C, 589 nm) (1 % in Methanol)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR