Tisk…

Beclomethasone dipropionate 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Beclometasone dipropionate

460990010 460992500
ACRO460990010EA 8830 CZK
ACRO460990010 ACRO460992500
Beclomethasone dipropionate 97%
Beclomethasone dipropionate
Vzorec: C₂₈H₃₇ClO₇
MW: 521.047 g/mol
Bod tání: 208 °C (rozkl.)
Storage Temperature: Refrigerator
MDL Number: MFCD00135613
CAS číslo: 5534-09-8

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C, 3 h)
HPLC ≥96.0 %
Specific optical rotation +88° to +94° (20°C, 589 nm) (1 % in dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR