Sulphasalazine 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
5-[4-(2-Pyridylsulphamoyl)phenylazo]salicylic Acid, 2-Hydroxy-5-((4-(N-(pyridin-2-yl)sulphamoyl)phenyl)diazenyl)benzoic acid
461240050
461240250
461241000
ACRO461240050EA
1430
CZK
ACRO461240050
ACRO461240250
ACRO461241000
Sulphasalazine 97%
Sulphasalazine
Vzorec:
C₁₈H₁₄N₄O₅S Bod tání: 260…265 °C Storage Temperature: Ambient |
MDL Number:
MFCD00057363 CAS číslo: 599-79-1 |
Výsledky specifikačního testu
Appearance | Light yellow to brownish-yellow Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % (1 g, 105°C, 2 h) |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...