Tisk…

Atracurium besylate (mixture of isomers) 96%

Dodavatel: Thermo Fisher Scientific

461470010 461470050
ACRO461470010EA 3140 CZK
ACRO461470010 ACRO461470050
Atracurium besylate (mixture of isomers) 96%
Atracurium besylate (mixture of isomers)
Vzorec: C₆₅H₈₂N₂O₁₈S₂
Bod tání: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00797403
CAS číslo: 64228-81-5

Objednat

Výsledky specifikačního testu

Appearance White to yellowish-white Powder
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥95.0 % (mix of isomers)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR