Daidzin 98%
Dodavatel: Thermo Scientific
Synonyms:
7-(beta-D-Glucopyranosyloxy)-4'-hydroxyisoflavone, 3-(4-Hydroxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one
460420010
460421000
ACRO460420010EA
17170
CZK
ACRO460420010
ACRO460421000
Daidzin 98%
Daidzin
Vzorec:
C₂₁H₂₀O₉ MW: 416,38 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00017466 CAS číslo: 552-66-9 |
Výsledky specifikačního testu
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2.5 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...