Tisk…

Cyproterone acetate 98%

Dodavatel: Thermo Fisher Scientific
Varování

459540010 459542500
ACRO459540010EA 8720 CZK
ACRO459540010 ACRO459542500
Cyproterone acetate 98%
Cyproterone acetate
Vzorec: C₂₄H₂₉ClO₄
MW: 416.94 g/mol
Bod tání: 207…215 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00864671
CAS číslo: 427-51-0

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 208 to 215 °C
Loss on drying ≤0.5 % (80 °C)
HPLC ≥97.5 %
Residue after ign. (sulfated) ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR