L-Theanine 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(S)-2-Amino-5-(ethylamino)-5-oxopentanoic acid, L-gamma-Glutamylethylamide, N-gamma-Ethyl-L-glutamine
459340010
459340050
ACRO459340010EA
1980
CZK
ACRO459340010
ACRO459340050
L-Theanine 98%
L-Theanine
Vzorec:
C₇H₁₄N₂O₃ MW: 174,20 g/mol Bod tání: 206…219 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00059653 CAS číslo: 3081-61-6 |
Výsledky specifikačního testu
Appearance | white to off-white Crystalline powder |
Infrared spectrum | Conforms |
Specific optical rotation | +7.5° to +8.5° (20°C, 589 nm) (1 % in water) |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...