Mycophenolic acid 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
459380050
459380250
459380010
ACRO459380050EA
3020
CZK
ACRO459380050
ACRO459380250
ACRO459380010
Mycophenolic acid 98%
Mycophenolic acid
Vzorec:
C₁₇H₂₀O₆ Storage Temperature: Refrigerator |
MDL Number:
MFCD00036814 CAS číslo: 24280-93-1 UN: 3077 ADR: 9,III |
Výsledky specifikačního testu
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 139 to 145 °C |
Loss on drying | ≤0.5 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...