(3-Indoxyl)-β-D-glucopyranoside 98%
Dodavatel: Thermo Scientific
Synonyms:
Indoxyl-D-glucoside, 3-Indole-D-glucoside, 3-Indolyl-D-glucoside, Indoxyl-B-D Glucoside, Indican (Plant Indican)
458880010
458882500
ACRO458880010EA
8170
CZK
ACRO458880010
ACRO458882500
(3-Indoxyl)-β-D-glucopyranoside 98%
(3-Indoxyl)-β-D-glucopyranoside
Vzorec:
C₁₄H₁₇NO₆ Bod tání: 57…58 °C Storage Temperature: Freezer |
MDL Number:
MFCD00047169 CAS číslo: 487-60-5 |
Výsledky specifikačního testu
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % |
HPLC | ≥97.5 % |
Specific optical rotation | -63.5° to -67.5° (20°C, 589 nm) (c=1, water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...