Tisk…

Tripropylene glycol (mixture of isomers) 99%, pure

Dodavatel: Thermo Scientific

Synonyms: Tripropylene glycol

458480010 458480050 458480250
ACRO458480010EA 1130 CZK
ACRO458480010 ACRO458480050 ACRO458480250
Tripropylene glycol (mixture of isomers) 99%, pure
Tripropylene glycol (mixture of isomers)
Vzorec: C₉H₂₀O₄
MW: 192,26 g/mol
Bod varu: 270 °C (760 mmHg)
Hustota: 1,02 g/cm³
Bod vzplanutí: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00014405
CAS číslo: 24800-44-0

Objednat

Výsledky specifikačního testu

Appearance Clear colorless Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4450 (20°C, 589 nm)
Water ≤0.05 %
GC ≥98.5 % (sum of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR