Tisk…

3-Acetylpyridine adenine dinucleotide 90%

Dodavatel: Thermo Fisher Scientific

457680010 457682500
ACRO457680010EA 34560 CZK
ACRO457680010 ACRO457682500
3-Acetylpyridine adenine dinucleotide 90%
3-Acetylpyridine adenine dinucleotide
Vzorec: C₂₂H₂₈N₆O₁₄P₂
Storage Temperature: Freezer
MDL Number: MFCD00078882
CAS číslo: 86-08-8

Objednat

Výsledky specifikačního testu

Appearance White to off-white to light yellow Powder
Infrared spectrum Conforms
Water ≤8 %
HPLC ≥89.0 %

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR