Tisk…

Proadifen hydrochloride 95%

Dodavatel: Thermo Scientific

Synonyms: Proadifen HCl

457880010 457880050
ACRO457880010EA 7320 CZK
ACRO457880010 ACRO457880050
Proadifen hydrochloride 95%
Proadifen hydrochloride
Vzorec: C₂₃H₃₁NO₂·HCl
MW: 389,964 g/mol
Bod tání: 122 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00055151
CAS číslo: 62-68-0

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Powder and/or chunks
Infrared spectrum Conforms
Melting point 121 to 123 °C
TLC ≥94 %
Chloride (Cl) 8.6 to 9.6 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR