Tisk…

Decyl decanoate 98%

Dodavatel: Thermo Scientific

457110250 457115000
ACRO457110250EA 1380 CZK
ACRO457110250 ACRO457115000
Decyl decanoate 98%
Decyl decanoate
Vzorec: C₂₀H₄₀O₂
MW: 312,5304 g/mol
Bod varu: 174 °C (2 mmHg)
Hustota: 0,859 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00059280
CAS číslo: 1654-86-0

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4430 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR