Inulin (from chicory root)
Dodavatel: Thermo Fisher Scientific
457100250
457101000
457105000
ACRO457100250EA
3870
CZK
ACRO457100250
ACRO457101000
ACRO457105000
Inulin (from chicory root)
Inulin
Storage Temperature:
Ambient |
MDL Number:
MFCD00131407 CAS číslo: 9005-80-5 |
Výsledky specifikačního testu
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤10 % (2 g, 105°C, 2 hrs) |
Free fructose, glucose,sucrose | ≤5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...