Azomethine H monosodium salt monohydrate 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4-Hydroxy-5-(2-hydroxybenzylideneamino)naphthalene-2,7-disulphonic acid monosodium salt
457090050
457090250
ACRO457090050EA
1610
CZK
ACRO457090050
ACRO457090250
Azomethine H monosodium salt monohydrate 95%
Azomethine H monosodium salt monohydrate
Vzorec:
C₁₇H₁₂NNaO₈S₂·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149588 CAS číslo: 304655-82-1 |
Výsledky specifikačního testu
Appearance | Yellow to orange Powder |
Infrared spectrum | Conforms |
Water | ≤6 % |
Titration with NaOH | ≥94 % |
Sodium (Na) | 4.5 to 5.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...