Tisk…

Copper(II) pyrithione

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Copper Omadine, Bis(1-Hydroxy-1H-pyridine-2-thionato-O,S)copper, Bis(2-pyridinethiol 1-oxide)copper

457210010 457210050
ACRO457210010EA 3210 CZK
ACRO457210010 ACRO457210050
Copper(II) pyrithione
Copper(II) pyrithione
Vzorec: C₁₀H₈CuN₂O₂S₂
MW: 315,85 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02660770
CAS číslo: 14915-37-8
UN: 2811
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance Green Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR