Cypermethrin 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
α-Cyano-3-phenoxybenzyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate
456310050
456310250
456311000
ACRO456310050EA
4930
CZK
ACRO456310050
ACRO456310250
ACRO456311000
Cypermethrin 98%
Cypermethrin
Vzorec:
C₂₂H₁₉Cl₂NO₃ MW: 416,3 g/mol Hustota: 1,12 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00055328 CAS číslo: 52315-07-8 UN: 2810 ADR: 6.1,III |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % (Mixture of isomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...