Lutein 90%
Dodavatel: Thermo Scientific
Synonyms:
E 161b, (6'R)-β,ε-carotene-3R,3'R-diol, Xanthophyll, (1R,4R)-4-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-18-((R)-4-Hydroxy-2,6,6-trimethylcyclohex-1-en-1-yl)-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaen-1-yl)-3,5,5-trimethylcyclohex-2-enol
456160010
456160050
ACRO456160010EA
9820
CZK
ACRO456160010
ACRO456160050
Lutein 90%
Lutein
Vzorec:
C₄₀H₅₆O₂ MW: 568,88 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD08435941 CAS číslo: 127-40-2 |
Výsledky specifikačního testu
Appearance | Yellow to red Powder |
Infrared spectrum | Conforms |
HPLC | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...