Tisk…

Imipenem monohydrate 98%

Dodavatel: Thermo Scientific

Synonyms: (5R,6S)-3-((2-Formimidamidoethyl)thio)-6-((R)-1-hydroxyethyl)-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid hydrate

455320010 455322500
ACRO455320010EA 22730 CZK
ACRO455320010 ACRO455322500
Imipenem monohydrate 98%
Imipenem monohydrate
Vzorec: C₁₂H₁₇N₃O₄S
MW: 317,36 g/mol
Bod tání: 193…198 °C
Storage Temperature: Freezer
MDL Number: MFCD09753321
CAS číslo: 74431-23-5

Objednat

Výsledky specifikačního testu

Appearance Off-white to light yellow Powder
Infrared spectrum Conforms
Water 5 to 8 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR