Phenyl vinyl sulphone 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
221250010
221250050
221250250
ACRO221250010EA
1080
CZK
ACRO221250010
ACRO221250050
ACRO221250250
Phenyl vinyl sulphone 98%
Phenyl vinyl sulphone
Vzorec:
C₈H₈O₂S Bod tání: 66…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007554 CAS číslo: 5535-48-8 |
Výsledky specifikačního testu
Appearance | White to beige Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 66 to 70 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...