Xanthosine dihydrate 99%
Dodavatel: Thermo Scientific
Synonyms:
9-β-D-Ribofuranosylxanthine dihydrate, 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purine-2,6-diol dihydrate, 3,9-Dihydro-9-beta-D-ribofuranosyl-1H-purine-2,6-dione dihydrate
207630010
207630050
ACRO207630010EA
1750
CZK
ACRO207630010
ACRO207630050
Xanthosine dihydrate 99%
Xanthosine dihydrate
Vzorec:
C₁₀H₁₂N₄O₆·2H₂O MW: 320,26 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00149347 CAS číslo: 5968-90-1 |
Výsledky specifikačního testu
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | 9 to 14 % |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...