Tisk…
Industrial gloves, ActivArmr® 97-681
Dodavatel: Ansell
97-681/11 97-681/8 97-681/9 97-681/10
ANSE97-681/11EA 3070 CZK
ANSE97-681/11 ANSE97-681/8 ANSE97-681/9 ANSE97-681/10
Industrial gloves, ActivArmr® 97-681
Rukavice
These nylon/acrylic gloves with full PVC coating provide mechanical and cold protection.
  • Leading dexterity among liquid-proof cold weather gloves
  • PVC foam coating (fully dipped) stays flexible down to −25 °C/−15 °F
  • Helps to fight frostbite
  • Industry leading combination of warmth and dexterity in a liquid-proof glove
RIPEL™ technology creates a liquid-impermeable layer in knit glove styles that prevents oil or lubricant from making even incidental contact with the wearer's skin, while also enhancing hand health.
Typical applications include: Cold weather protection, drilling/extraction, infrastructure, production services.
Certifikace: EN 388:2016 (2231B), EN 511 (221); Cat. II
Food contact approved