Tisk…
Doxorubicin hydrochloride ≥98%
Dodavatel: Cayman Chemical
Synonyms: Doxorubicin HCl, Hydroxydaunorubicin Hydrochloride, (8S,10S)-10-(((2R,4S,5S,6S)-4-Amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride
15007-5 15007-10 15007-25 15007-50
CAYM15007-5EA 1320 CZK
CAYM15007-5 CAYM15007-10 CAYM15007-25 CAYM15007-50
Doxorubicin hydrochloride ≥98%
Doxorubicin hydrochloride
Doxorubicin is an anthracycline anti-tumour antibiotic which involves in inhibition of DNA topoisomerase II by inducing double-stranded DNA breaks. It is also involved in the inhibition of nucleic acid synthesis by intercalating within DNA. The accumulation of the p53 tumour suppressor protein leads to the apoptosis in cells.
  • Crystalline solid
Vzorec: C₂₇H₂₉NO₁₁·HCl
MW: 579,98 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077757
CAS číslo: 25316-40-9