Tisk…

N-trans-Cinnamyl-N-methyl-(1-naphthylmethyl)amine Hydrochloride 99%

Dodavatel: Thermo Fisher Scientific

Synonyms: Naftifine HCl, (E)-N-methyl-N-(naphthalen-1-ylmethyl)-3-phenylprop-2-en-1-amine hydrochloride, AW 105843, SN 105843, N-trans-Cinnamyl-N-methyl-(1-naphthylmethyl)amine hydrochloride

458660010 458660050 458660250
ACRO458660010EA 2350 CZK
ACRO458660010 ACRO458660050 ACRO458660250
N-trans-Cinnamyl-N-methyl-(1-naphthylmethyl)amine Hydrochloride 99%
Naftifine hydrochloride
Vzorec: C₂₁H₂₁N
Bod tání: 175…179 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00059047
CAS číslo: 65473-14-5

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 174 to 180 °C
Titration with HClO4 ≥98.5 %
Loss on fusion ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR