Tisk…

Ofloxacin 98%

Dodavatel: Thermo Scientific

Synonyms: 9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid, 9-Fluoro-3,7-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid

455670010 455670050 455670250
ACRO455670010EA 1660 CZK
ACRO455670010 ACRO455670050 ACRO455670250
Ofloxacin 98%
Ofloxacin
Vzorec: C₁₈H₂₀FN₃O₄
MW: 361,37 g/mol
Bod tání: 280…275 °C
Storage Temperature: Freezer
MDL Number: MFCD00226105
CAS číslo: 82419-36-1

Objednat

Výsledky specifikačního testu

Appearance White to yellow Powder
Infrared spectrum Conforms
Loss on drying ≤0.2 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR