Tisk…

Oxacillin sodium salt 95%

Dodavatel: Thermo Scientific
Nebezpečí

455440010 455440050
ACRO455440010EA 1410 CZK
ACRO455440010 ACRO455440050
Oxacillin sodium salt 95%
Oxacillin sodium salt
Vzorec: C₁₉H₁₈N₃NaO₅S
MW: 423,42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00056864
CAS číslo: 1173-88-2

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water 3.5 to 5.0 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR