Chromotrope 2B, pure
Dodavatel: Thermo Fisher Scientific
|
Varování
|
149750050
149750250
ACRO149750050EA
912
CZK
ACRO149750050
ACRO149750250
Chromotrope 2B, pure
Chromotrope 2B
Vzorec:
C₁₆H₉N₃Na₂O₁₀S₂ Bod tání: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003951 CAS číslo: 548-80-1 |
Výsledky specifikačního testu
Appearance | Dark green to greenish-brown to brown Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤18 % |
UV Lambda max | 513 to 517 nm (c=0.1, H2O) |
Specific extinction E (1%/1cm) | ≥580 (on dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...