Tisk…
Antimony standard solution, for ICP, in 30% HCl, VHG
Dodavatel: LGC Standards PROMOCHEM
VHG-PSBH-100 VHG-TSBH-100 VHG-PSBH-50 VHG-PSBH-500 VHG-TSBH-500
PROOVHG-PSBH-100EA 1710 CZK
PROOVHG-PSBH-100 PROOVHG-TSBH-100 PROOVHG-PSBH-50 PROOVHG-PSBH-500 PROOVHG-TSBH-500
Antimony standard solution, for ICP, in 30% HCl, VHG
Standardy Standardy kationické Standardy - antimon
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifikace: Traceable to NIST, SRM.