Tisk…
Platinum standard solution, for ICP, in 20% HCl, VHG
Dodavatel: LGC Standards PROMOCHEM
VHG-PPTH-100 VHG-PPTH-50 VHG-PPTH-500 VHG-TPTH-100 VHG-TPTH-500
PROOVHG-PPTH-100EA 2500 CZK
PROOVHG-PPTH-100 PROOVHG-PPTH-50 PROOVHG-PPTH-500 PROOVHG-TPTH-100 PROOVHG-TPTH-500
Platinum standard solution, for ICP, in 20% HCl, VHG
Standardy Standardy kationické Standardy - platina
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifikace: Traceable to NIST, SRM.