Tisk…
Galantamine Hydrobromide, Reference Standards
Dodavatel: USP
1287755 1287766 1287777
1287755.EA 15170 CZK
1287755. 1287766. 1287777.
Galantamine Hydrobromide, Reference Standards
Standardy Standardy organické
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifikace: US Pharmacopea Reference Standard
Objednat
Produktová specifikace
Popis Galantamine Hydrobromide Galantamine Hydrobromide Racemic Galantamine Hydrobromide Related Compounds Mixture
Environmentally Preferable
Čí. CAS 1953-04-4; 69353-21-5 193146-85-9
Vzorec C₁₇H₂₂BrNO₃ C₁₇H₂₂BrNO₃
Č. UN 1544 1544 1544