Tisk…

Calcium-D-saccharate tetrahydrate 98%

Dodavatel: Thermo Fisher Scientific
Varování

J66947.14 J66947.22
J66947.14EA 2780 CZK
J66947.14 J66947.22
Calcium-D-saccharate tetrahydrate 98%
Calcium-D-saccharate tetrahydrate

D-Saccharate (D-glucaric acid) is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.


  • Soluble in water( 0,43 g/l)


Upozornení: Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Vzorec: C₆H₈CaO₈.4H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150704
CAS číslo: 5793-89-5
EINECS: 227-334-1
Merck Index: 14,1700

Objednat

Výsledky specifikačního testu

Colour White
Form Powder

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR