Tisk…

Ethylene glycol monovinyl ether ≥96.5% (by GC), Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Vinyl Cellosolve, Ethylene glycol vinyl ether, 2-Vinyloxyethanol, Vinyl Glycol, 2-(Vinyloxy)ethan-1-ol

M-Clarity™ quality level = MQ200

410020-10ML 410020-50ML
SIAL410020-10MLEA 1370 CZK
SIAL410020-10ML SIAL410020-50ML
Ethylene glycol monovinyl ether ≥96.5% (by GC), Sigma-Aldrich®
Ethylene glycol monovinyl ether
Vzorec: C₄H₈O₂
MW: 88,11 g/mol
Bod varu: 143 °C (lit.)
Hustota: 0,982 g/mL (25 °C) (lit.)
MDL Number: MFCD00192151
CAS číslo: 764-48-7
UN: 3271
ADR: 3,III

Objednat

Výsledky specifikačního testu

Purity (GC) 96,5%
Refractive index (n20/D) 1,436 (lit.)
Form Liquid
Colour Colourless
Infrared spectrum Conforms to structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR