Tisk…

Aluminum silicate, pearls, orange desiccant heavy metal-free, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Aluminosilicate, Drying Pearls, Orange, Silica Gel Orange, Vitreous Aluminosilicate

94098-1KG 94098-500G
SIAL94098-1KGEA 2830 CZK
SIAL94098-1KG SIAL94098-500G
Aluminum silicate, pearls, orange desiccant heavy metal-free, Sigma-Aldrich®
Aluminum silicate

Drying pearls changes colour from orange to colorless after moisture absorption.


regeneration by drying at 130 - 160 °C; do not heat above 160 °C.

Vzorec: Al₂O₅Si
MDL Number: MFCD03098976
CAS číslo: 1327-36-2

Objednat

Výsledky specifikačního testu

Absorption Capacity (Water) >30%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR