Ethanol eurodenatured 96, TechniSolv® (1:1:1)
Dodavatel: VWR Chemicals
|
|
85824.360EA
2130
CZK
85824.360
85824.440
Ethanol eurodenatured 96, TechniSolv® (1:1:1)
Ethanol Eurodenatured
Vzorec:
H₃CCH₂OH MW: 46.07 g/mol |
CAS číslo:
64-17-5 UN: 1170 ADR: 3 II |
Výsledky specifikačního testu
Assay (V/V) | min. 93.8% |
Denatonium benzoate (ppm) | 11.8 - 13.8 ppm |
Methyl ethyl ketone (V/V) | 0.93 - 1.09 % |
2-Propanol (V/V) | 0.93 - 1.09% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...