Tisk…
Amiodarone, Reference Standards
Dodavatel: USP
1027302 1027346 1027357 1027380
1027302.EA 10580 CZK
1027302. 1027346. 1027357. 1027380.
Amiodarone, Reference Standards
Standardy Standardy organické
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifikace: US Pharmacopea Reference Standard
Objednat
Produktová specifikace
Popis Amiodarone Hydrochloride Amiodarone Related Compound D Amiodarone Related Compound E Amiodarone Related Compound H
Synonym(s) (2-Butylbenzofuran-3-yl)(4-hydroxy-3,5-diiodophenyl)methanone (2-butylbenzofuran-3-yl)(4-hydroxyphenyl)methanone 2-chloro-N,N-diethylethanamine hydrochloride
Environmentally Preferable
Čí. CAS 19774-82-4 1951-26-4 52490-15-0 869-24-9
Vzorec C₂₅H₃₀ClI₂NO₃ C₁₉H₁₆I₂O₃ C₁₉H₁₈O₃ C₆H₁₅Cl₂N
Č. UN 3077 2928