Tisk…
Dihydrogen hexahydroxyplatinate(IV) ≥99.9% (trace metals basis), Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Hydrogen hexahydroxyplatinate(IV)
M-Clarity™ quality level = MQ100
334472-2.5G 334472-500MG
SIAL334472-2.5GEA 19030 CZK
SIAL334472-2.5G SIAL334472-500MG
Dihydrogen hexahydroxyplatinate(IV) ≥99.9% (trace metals basis), Sigma-Aldrich®
Dihydrogen hexahydroxyplatinate(IV)
Synonym: Platinic acid.
Vzorec: H₂Pt(OH)₆
MW: 299,14 g/mol
MDL Number: MFCD00058744
CAS číslo: 51850-20-5
Objednat
Výsledky specifikačního testu
Purity (based on trace metal analysis) 99,9%
Trace metal analysis <1500,0 ppm
Platinum (Pt) 58,9 - 71,7 %
Form Powder or crystal or chunk
Colour Yellow
Solubility H₂O, aqueous acid (slightly soluble)