(±)-Alliin ≥98%, crystalline solid
Dodavatel: Cayman Chemical
Alliin is a cysteine sulfoxide constituent of garlic that is converted by alliinase to allicin, which imparts its pungent aroma and flavor.
- A crystalline solid
Alliin exhibits anti-cancer, anti-microbial, anti-hypertensive, cardioprotective, and anti-oxidant activities. It has been shown to inhibit squalene monooxygenase, a rate-limiting step in the downstream pathway of cholesterol biosynthesis, with an IC50 value of 120 μM.
Storage Temperature:
Freezer |
CAS číslo:
17795-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...