Tisk…

Lamotrigine ≥98%

Dodavatel: Cayman Chemical

Synonyms: 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, 3,5-Diamino-6-(2,3-dichlorophenyl)-1,2,4-triazine

15428-10 15428-25 15428-100 15428-50
CAYM15428-10EA 2330 CZK
CAYM15428-10 CAYM15428-25 CAYM15428-100 CAYM15428-50
Lamotrigine ≥98%
Lamotrigine

Lamotrigine is an inhibitor of voltage-gated sodium channels (Nav), blocking human Nav1,2, Nav1,5, and Nav1,8 with IC50 values of 10, 62, and 96 μM, respectively.


Presumably through this action, lamotrigine acts as an anticonvulsant and has applications in many diseases that feature seizures, including epilepsy and autism spectrum disorder.

Vzorec: C₉H₇Cl₂N₅
Storage Temperature: Freezer
MDL Number: MFCD00865333
CAS číslo: 84057-84-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR