Hydroxynaphthol blue indicator for metal titration, Fluka™
Dodavatel: Honeywell Chemicals
Synonyms:
1-(2-Hydroxy-4-sulpho-1-naphthylazo)-2-naphthol-3,6-disulphonic acid trisodium salt
33936-10G
33936-50G
33936-10GEA
3050
CZK
33936-10G
33936-50G
Hydroxynaphthol blue indicator for metal titration, Fluka™
Hydroxynaphthol blue
Vzorec:
C₂₀H₁₁N₂Na₃O₁₁S₃ MW: 620,47 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00036393 CAS číslo: 63451-35-4 EINECS: 264-197-7 |
Výsledky specifikačního testu
Identity (LC-MS) | complying |
Absorption maximum | 640 - 650 nm |
Absorbtion (1%/1cm) | Min. 325 |
Suitability for Complexometry | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...