Tisk…
Septum stoppers, rubber, Suba-Seal®, Sigma-Aldrich®
Dodavatel: Merck
Z124591-100EA Z124680-100EA
SIALZ124591-100EAEA 7400 CZK
SIALZ124591-100EA SIALZ124680-100EA
Septum stoppers, rubber, Suba-Seal®, Sigma-Aldrich®
Zátky Zátky kaučukové (gumové)
These stoppers are ideal for air and moisture-sensitive chemistry procedures. After piercing with a non-coring needle, the rubber closes the puncture, providing an air and moisture-tight seal to protect contents of vessel or container from atmosphere. These closures have a turnover flange molded to grip the outside of the container neck promoting a double seal in conjunction with annular serrations on hollow plug.